siRNA Details
| Gene Symbol: Ttll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133923.6 | NP_598684.4 | 13 | 2484 | ||
| GenBank mRNA | AK080321.1 | - | 9 | 1898 | ||
| AM690746.1 | - | 13 | 2484 | |||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Ttll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133923.6 | NP_598684.4 | 13 | 2484 | ||
| GenBank mRNA | AK080321.1 | - | 9 | 1898 | ||
| AM690746.1 | - | 13 | 2484 | |||
Ttll3
Genomic Map