siRNA Details
| Gene Symbol: Snx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133924.3 | NP_598685.2 | 3 | 543 | ||
| GenBank mRNA | AK141663.1 | - | 3 | 544 | ||
| AK161207.1 | - | 1 | 142 | |||
| AK168473.1 | - | 3 | 530 | |||
| BC094371.1 | - | 2 | 1239 | |||
| BC117542.1 | - | 4 | 763 | 40105600 | ||

Snx21
Genomic Map