siRNA Details
| Gene Symbol: Fbxl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133940.3 | NP_598701.2 | 1 | 669 | ||
| GenBank mRNA | AK036124.1 | - | 1 | 565 | ||
| AK084506.1 | - | 1 | 556 | |||
| AK155154.1 | - | 1 | 552 | |||
| BC006913.1 | - | 1 | 258 | |||
| BC021329.1 | - | 1 | 562 | 3710945 | ||

Fbxl14
Genomic Map