siRNA Details
| Gene Symbol: AI467606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178901.3 | NP_849232.1 | 2 | 583 | ||
| GenBank mRNA | AK041954.1 | - | 2 | 639 | ||
| AK052739.1 | - | 2 | 584 | |||
| AK155213.1 | - | 2 | 638 | |||
| AK168260.1 | - | 2 | 585 | |||
| AK169672.1 | - | 2 | 662 | |||
| AK170238.1 | - | 2 | 595 | |||
| BC038694.1 | - | 2 | 569 | 1513939 | ||
| BC064101.1 | - | 2 | 570 | 1366571 | ||

AI467606
Genomic Map