siRNA Details
| Gene Symbol: Ano1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242349.1 | NP_001229278.1 | 17 | 2052 | ||
| NM_178642.5 | NP_848757.4 | 18 | 2061 | |||
| GenBank mRNA | AK029329.1 | - | 4 | 428 | ||
| AK032793.1 | - | 13 | 1073 | |||
| AK052589.1 | - | 18 | 2013 | |||
| BC004697.1 | - | 10 | 927 | |||
| BC062959.1 | - | 17 | 1938 | 30547439 | ||

Ano1
Genomic Map