siRNA Details
| Gene Symbol: Spred3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182927.3 | NP_891557.3 | 6 | 1260 | ||
| GenBank mRNA | AB115762.1 | - | 5 | 1128 | ||
| AK051617.1 | - | 3 | 760 | |||
| BC052757.1 | - | 6 | 1080 | |||
| BC062119.1 | - | 6 | 1260 | 6821036 | ||

Spred3
Genomic Map