siRNA Details
| Gene Symbol: AW146154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033530.3 | NP_001028702.1 | 2 | 708 | ||
| GenBank mRNA | AK164198.1 | - | 2 | 709 | ||
| BC138895.1 | - | 2 | 466 | 8861917 | ||
| BC138896.1 | - | 2 | 466 | 8861918 | ||

AW146154
Genomic Map