siRNA Details
| Gene Symbol: Plekhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081333.1 | NP_001074802.1 | 14 | 2461 | ||
| GenBank mRNA | AK029859.1 | - | 12 | 3133 | ||
| AK131125.1 | - | 10 | 3428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhg4| Gene Symbol: Plekhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081333.1 | NP_001074802.1 | 14 | 2461 | ||
| GenBank mRNA | AK029859.1 | - | 12 | 3133 | ||
| AK131125.1 | - | 10 | 3428 | |||