siRNA Details
| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133962.3 | NP_598723.3 | 5 | 535 | ||
| GenBank mRNA | AK041423.1 | - | 5 | 538 | ||
| AK147923.1 | - | 5 | 506 | |||
| BC060639.1 | - | 5 | 507 | 6849875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef18| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133962.3 | NP_598723.3 | 5 | 535 | ||
| GenBank mRNA | AK041423.1 | - | 5 | 538 | ||
| AK147923.1 | - | 5 | 506 | |||
| BC060639.1 | - | 5 | 507 | 6849875 | ||