siRNA Details
Gene Symbol: Enkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198299.1 | NP_938041.1 | 7 | 1009 | ||
GenBank mRNA | AK087475.1 | - | 6 | 1172 | ||
BC018168.1 | - | 6 | 1295 | |||
BC023921.1 | - | 6 | 1398 | |||
BC026993.1 | - | 4 | 3076 | |||
BC052498.1 | - | 7 | 1009 | 1245862 |
