siRNA Details
| Gene Symbol: Cmtm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133978.2 | NP_598739.1 | 2, 3 | 504 | ||
| GenBank mRNA | AF188504.1 | - | 2, 3 | 340 | ||
| AK151785.1 | - | 2, 3 | 307 | |||
| AK167886.1 | - | 2, 3 | 350 | |||
| AK168024.1 | - | 2, 3 | 357 | |||
| AY241869.1 | - | 2, 3 | 361 | |||

Cmtm7
Genomic Map