siRNA Details
| Gene Symbol: Cmtm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133978.2 | NP_598739.1 | 3 | 556 | ||
| GenBank mRNA | AF188504.1 | - | 3 | 392 | ||
| AK040123.1 | - | 2 | 1633 | |||
| AK151785.1 | - | 3 | 359 | |||
| AK167886.1 | - | 3 | 402 | |||
| AK168024.1 | - | 3 | 409 | |||
| AK168280.1 | - | 3 | 393 | |||
| AK215113.1 | - | 1 | 66 | |||
| AY241869.1 | - | 3 | 413 | |||

Cmtm7
Genomic Map