siRNA Details
| Gene Symbol: Cmtm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252479.1 | NP_001239408.1 | 3 | 544 | ||
| NM_133978.2 | NP_598739.1 | 4 | 643 | |||
| GenBank mRNA | AF188504.1 | - | 4 | 479 | ||
| AK151785.1 | - | 4 | 446 | |||
| AK167886.1 | - | 4 | 489 | |||
| AK168024.1 | - | 4 | 496 | |||
| AK168280.1 | - | 4 | 480 | |||
| AY241869.1 | - | 4 | 500 | |||
| BC038633.1 | - | 3 | 383 | 1066813 | ||

Cmtm7
Genomic Map