siRNA Details
| Gene Symbol: Slc6a20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139142.2 | NP_631881.1 | 5 | 697 | ||
| GenBank mRNA | AJ428067.1 | - | 710 | |||
| AJ964961.1 | - | 5 | 671 | |||
| AK043297.1 | - | 5 | 708 | |||
| AK080979.1 | - | 708 | ||||
| AK140445.1 | - | 708 | ||||
| AK160468.1 | - | 708 | ||||
| BC118933.2 | - | 5 | 697 | 40090800 | ||

Slc6a20a
Genomic Map