siRNA Details
| Gene Symbol: Tcta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133986.1 | NP_598747.1 | 1 | 133 | ||
| GenBank mRNA | AK133663.1 | - | 172 | |||
| BC019397.1 | - | 1 | 133 | 3672301 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcta| Gene Symbol: Tcta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133986.1 | NP_598747.1 | 1 | 133 | ||
| GenBank mRNA | AK133663.1 | - | 172 | |||
| BC019397.1 | - | 1 | 133 | 3672301 | ||