siRNA Details
| Gene Symbol: B630019K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046767.1 | - | 1 | 395 | ||
| BC055780.1 | - | 1 | 316 | 6403722 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB659663.1 | - | 146 | Chr. - - [-] on Build | ||

B630019K06Rik
Genomic Map