siRNA Details
| Gene Symbol: AU022751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033211.3 | NP_001028383.2 | 2 | 615 | ||
| NM_001166433.1 | NP_001159905.1 | 1 | 978 | |||
| GenBank mRNA | AK087702.1 | - | 1 | 977 | ||
| AK136005.1 | - | 2 | 616 | |||
| AK166653.1 | - | 2 | 615 | |||
| BC100377.1 | - | 1 | 964 | 30938251 | ||

AU022751
Genomic Map