siRNA Details
| Gene Symbol: Fig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133999.1 | NP_598760.1 | 14, 15 | 1776 | ||
| GenBank mRNA | AK041196.1 | - | 14, 15 | 1777 | ||
| AK172130.1 | - | 14, 15 | 1770 | |||
| AK172341.1 | - | 14, 15 | 1741 | |||
| AK172926.1 | - | 7, 8 | 824 | |||
| AK181110.1 | - | 3, 4 | 240 | |||
| AK204518.1 | - | 3, 4 | 305 | |||
| BC015295.1 | - | 14, 15 | 1776 | 4221630 | ||
| BC031887.1 | - | 14, 15 | 1775 | 5363304 | ||

Fig4
Genomic Map