siRNA Details
| Gene Symbol: Tmem263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013028.2 | NP_001013046.1 | 3 | 666 | ||
| GenBank mRNA | AK005698.1 | - | 551 | |||
| AK142035.1 | - | 3 | 519 | |||
| BC086492.1 | - | 3 | 531 | 30470653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem263| Gene Symbol: Tmem263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013028.2 | NP_001013046.1 | 3 | 666 | ||
| GenBank mRNA | AK005698.1 | - | 551 | |||
| AK142035.1 | - | 3 | 519 | |||
| BC086492.1 | - | 3 | 531 | 30470653 | ||