siRNA Details
| Gene Symbol: Zfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034895.3 | NP_001030067.2 | 12, 13 | 1929 | ||
| GenBank mRNA | AK033649.1 | - | 5, 6 | 678 | ||
| AK122438.1 | - | 5, 6 | 3537 | |||
| BC072571.1 | - | 11, 12 | 2598 | |||
| BF449481.1 | - | 2, 3 | 304 | |||

Zfr2
Genomic Map