siRNA Details
| Gene Symbol: Zfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034895.3 | NP_001030067.2 | 13 | 2047 | ||
| GenBank mRNA | AK033649.1 | - | 6 | 796 | ||
| AK122438.1 | - | 6 | 3655 | |||
| BC072571.1 | - | 12 | 2716 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfr2| Gene Symbol: Zfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034895.3 | NP_001030067.2 | 13 | 2047 | ||
| GenBank mRNA | AK033649.1 | - | 6 | 796 | ||
| AK122438.1 | - | 6 | 3655 | |||
| BC072571.1 | - | 12 | 2716 | |||