siRNA Details
Gene Symbol: Ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134009.3 | NP_598770.1 | 2, 3 | 469 | ||
GenBank mRNA | AK019409.1 | - | 2, 3 | 339 | ||
AK048954.1 | - | 2, 3 | 463 | |||
AK128887.1 | - | 2, 3 | 470 | |||
AK177319.1 | - | 1, 2 | 30 | |||
BC019501.1 | - | 2, 3 | 469 | 4193809 |
