siRNA Details
| Gene Symbol: Mbtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134012.3 | NP_598773.2 | 7 | 929 | ||
| GenBank mRNA | AK033238.1 | - | 2 | 3802 | ||
| AK141155.1 | - | 7 | 924 | |||
| AK153919.1 | - | 9 | 764 | |||
| BC020018.1 | - | 3 | 240 | 2646754 | ||
| BC062907.1 | - | 7 | 929 | 6813849 | ||
| BC064014.1 | - | 9 | 1033 | 6335954 | ||

Mbtd1
Genomic Map