siRNA Details
| Gene Symbol: Smg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002764.1 | NP_001002764.1 | 2 | 328 | ||
| GenBank mRNA | BC016550.1 | - | 2 | 281 | ||
| BC047279.1 | - | 2 | 339 | |||
| BC066040.1 | - | 2 | 328 | 6410245 | ||
| BC077715.1 | - | 2 | 291 | |||

Smg6
Genomic Map