siRNA Details
| Gene Symbol: 6330403K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134022.2 | NP_598783.1 | 1 | 566 | ||
| GenBank mRNA | AJ271140.1 | - | 1 | 404 | ||
| AK018106.1 | - | 1 | 463 | |||
| AK083923.1 | - | 1 | 411 | |||
| BC069976.1 | - | 1 | 404 | 6518302 | ||

6330403K07Rik
Genomic Map