siRNA Details
| Gene Symbol: Mien1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025559.2 | NP_079835.1 | 1, 2 | 121 | ||
| GenBank mRNA | AK002959.1 | - | 1, 2 | 104 | ||
| AK007795.1 | - | 1, 2 | 104 | |||
| AK009922.1 | - | 1, 2 | 99 | |||
| AK041314.1 | - | 1, 2 | 103 | |||
| BC021589.1 | - | 1, 2 | 105 | 5100542 | ||
| BE623929.1 | - | 1, 2 | 121 | |||

Mien1
Genomic Map