siRNA Details
| Gene Symbol: Slc25a41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175333.3 | NP_780542.1 | 2 | 105 | ||
| GenBank mRNA | AK077111.1 | - | 2 | 106 | ||
| BC115588.1 | - | 3 | 204 | 40102382 | ||
| BC115589.1 | - | 2 | 98 | 40102384 | ||
| FM165286.1 | - | 2 | 47 | |||

Slc25a41
Genomic Map