siRNA Details
| Gene Symbol: Zfp692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040686.1 | NP_001035776.1 | 12 | 1585 | ||
| NM_182996.3 | NP_892041.2 | 12 | 1647 | |||
| GenBank mRNA | AK031177.1 | - | 11 | 1875 | ||
| AK154896.1 | - | 1589 | ||||
| AK170229.1 | - | 11 | 1938 | |||
| AK170358.1 | - | 11 | 1478 | |||
| BC044879.1 | - | 6 | 1289 | |||
| BC055041.1 | - | 10 | 1984 | |||
| BC145470.1 | - | 12 | 1544 | 9054032 | ||
| BC150776.1 | - | 12 | 1544 | 9087687 | ||

Zfp692
Genomic Map