siRNA Details
| Gene Symbol: Hoxb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134032.2 | NP_598793.2 | 2 | 1195 | ||
| GenBank mRNA | AB221540.1 | - | 1 | 441 | ||
| BC019414.1 | - | 1 | 780 | |||
| BC138494.1 | - | 2 | 1114 | 8861514 | ||
| BC138495.1 | - | 2 | 1114 | 8861515 | ||

Hoxb2
Genomic Map