siRNA Details
| Gene Symbol: Plin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113471.1 | NP_001106942.1 | 8 | 1217 | ||
| NM_175640.2 | NP_783571.2 | 8 | 1273 | |||
| GenBank mRNA | AK031445.1 | - | 7 | 1198 | ||
| AY161165.1 | - | 7 | 1168 | |||
| BC096685.1 | - | 8 | 1218 | 1448913 | ||

Plin1
Genomic Map