siRNA Details
| Gene Symbol: Synj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045515.1 | NP_001038980.1 | 22, 23 | 3013 | ||
| NM_001164483.1 | NP_001157955.1 | 22, 23 | 3106 | |||
| GenBank mRNA | AB093272.1 | - | 21, 22 | 2899 | ||
| BU703932.1 | - | 1, 2 | 56 | |||
| BZ690371.2 | - | 2, 3 | 229 | |||
| CF741221.1 | - | 4, 5 | 495 | |||

Synj1
Genomic Map