siRNA Details
| Gene Symbol: Synj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045515.1 | NP_001038980.1 | 5 | 722 | ||
| NM_001164483.1 | NP_001157955.1 | 5 | 815 | |||
| GenBank mRNA | AB093272.1 | - | 4 | 608 | ||
| AK080946.1 | - | 5 | 689 | |||

Synj1
Genomic Map