siRNA Details
| Gene Symbol: Synpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109975.1 | NP_001103445.1 | 2 | 810 | ||
| NM_177340.2 | NP_796314.2 | 2 | 751 | |||
| GenBank mRNA | AK129267.1 | - | 2 | 701 | ||
| AK141564.1 | - | 2 | 812 | |||
| AK154958.1 | - | 2 | 746 | |||
| AK158933.1 | - | 3 | 774 | |||
| BC065127.1 | - | 2 | 1017 | |||

Synpo
Genomic Map