siRNA Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019435.4 | NP_062308.2 | 2 | 543 | ||
| GenBank mRNA | AK028121.1 | - | 2 | 264 | ||
| BC096047.1 | - | 2 | 239 | |||
| Y08702.1 | - | 2 | 236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufb11| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019435.4 | NP_062308.2 | 2 | 543 | ||
| GenBank mRNA | AK028121.1 | - | 2 | 264 | ||
| BC096047.1 | - | 2 | 239 | |||
| Y08702.1 | - | 2 | 236 | |||