siRNA Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019435.4 | NP_062308.2 | 3 | 740 | ||
| GenBank mRNA | AK028121.1 | - | 3 | 461 | ||
| AV086840.1 | - | 1 | 119 | |||
| BC027265.1 | - | 3 | 733 | 3589704 | ||
| BC096047.1 | - | 3 | 436 | |||
| Y08702.1 | - | 3 | 433 | |||

Ndufb11
Genomic Map