siRNA Details
| Gene Symbol: Etv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023794.2 | NP_076283.2 | 13 | 1534 | ||
| GenBank mRNA | AK018368.1 | - | 13 | 1586 | ||
| AK031452.1 | - | 13 | 1581 | |||
| AK049234.1 | - | 13 | 1584 | |||
| AK080674.1 | - | 13 | 1511 | |||
| AK164091.1 | - | 13 | 1584 | |||
| AK168881.1 | - | 13 | 1582 | |||
| AK205269.1 | - | 1 | 26 | |||
| AY004174.1 | - | 12 | 1411 | |||
| BC034680.1 | - | 13 | 1534 | 4036564 | ||

Etv5
Genomic Map