siRNA Details
| Gene Symbol: Etv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023794.2 | NP_076283.2 | 4 | 292 | ||
| GenBank mRNA | AK018368.1 | - | 4 | 344 | ||
| AK031452.1 | - | 4 | 339 | |||
| AK049234.1 | - | 4 | 341 | |||
| AK080674.1 | - | 3 | 215 | |||
| AK164091.1 | - | 4 | 341 | |||
| AK168881.1 | - | 4 | 340 | |||
| AY004174.1 | - | 3 | 169 | |||
| BC034680.1 | - | 4 | 292 | 4036564 | ||

Etv5
Genomic Map