siRNA Details
| Gene Symbol: Rhoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145491.2 | NP_663466.2 | 3 | 740 | ||
| GenBank mRNA | AB060650.1 | - | 3 | 558 | ||
| BC048813.1 | - | 3 | 550 | 2582017 | ||
| BC056363.1 | - | 3 | 740 | 6821501 | ||
| BC057451.1 | - | 3 | 272 | |||

Rhoq
Genomic Map