siRNA Details
| Gene Symbol: Cabin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172549.3 | NP_766137.2 | 34 | 6267 | ||
| GenBank mRNA | AK039606.1 | - | 8 | 1970 | ||
| AK078480.1 | - | 6 | 1186 | |||
| AK169961.1 | - | 6 | 1124 | |||
| AK171567.1 | - | 6 | 1152 | |||
| BC030927.1 | - | 5 | 1017 | |||
| BC056651.1 | - | 3 | 474 | |||
| BC150825.1 | - | 33 | 5831 | 9087736 | ||

Cabin1
Genomic Map