siRNA Details
| Gene Symbol: Tex15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031374.2 | NP_113551.2 | 1 | 2254 | ||
| GenBank mRNA | AF285589.1 | - | 1 | 2254 | ||
| AK143304.1 | - | 6 | 3026 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex15| Gene Symbol: Tex15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031374.2 | NP_113551.2 | 1 | 2254 | ||
| GenBank mRNA | AF285589.1 | - | 1 | 2254 | ||
| AK143304.1 | - | 6 | 3026 | |||