siRNA Details
| Gene Symbol: Isl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027397.3 | NP_081673.2 | 3 | 509 | ||
| GenBank mRNA | AK013964.1 | - | 3 | 452 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Isl2| Gene Symbol: Isl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027397.3 | NP_081673.2 | 3 | 509 | ||
| GenBank mRNA | AK013964.1 | - | 3 | 452 | ||