siRNA Details
| Gene Symbol: Meig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008579.4 | NP_032605.1 | 2 | 299 | ||
| GenBank mRNA | AK006347.1 | - | 2 | 113 | ||
| AK007239.1 | - | 2 | 113 | |||
| BC049538.1 | - | 2 | 299 | 6773052 | ||
| X64455.1 | - | 2 | 109 | |||

Meig1
Genomic Map