siRNA Details
| Gene Symbol: Rhox9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023894.1 | NP_076383.1 | 2 | 585 | ||
| GenBank mRNA | AF201698.1 | - | 2 | 585 | ||
| AK018818.1 | - | 1 | 26 | |||
| BC099451.1 | - | 2 | 524 | 30716051 | ||
| BC099963.1 | - | 2 | 519 | 30083999 | ||
| DQ058646.1 | - | 2 | 585 | |||

Rhox9
Genomic Map