siRNA Details
| Gene Symbol: 0610010K14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177601.1 | NP_001171072.1 | 5 | 561 | ||
| NM_026757.2 | NP_081033.1 | 4 | 459 | |||
| NM_145758.1 | NP_665701.1 | 5 | 561 | |||
| GenBank mRNA | AA189586.1 | - | 2 | 67 | ||
| AK003842.1 | - | 4 | 459 | |||
| AK178853.1 | - | 279 | ||||
| AK208696.1 | - | 289 | ||||
| CK791552.1 | - | 5 | 540 | |||
| EL959640.1 | - | 5 | 506 | |||

0610010K14Rik
Genomic Map