siRNA Details
| Gene Symbol: Mycbpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170671.2 | NP_733771.2 | 14, 15 | 2193 | ||
| GenBank mRNA | AK029929.1 | - | 13, 14 | 2369 | ||
| AK166780.1 | - | 7, 8 | 1341 | |||
| BC027327.1 | - | 10, 11 | 1554 | 3991131 | ||
| BC050808.1 | - | 3, 4 | 716 | |||

Mycbpap
Genomic Map