siRNA Details
| Gene Symbol: Ttc7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033213.1 | NP_001028385.1 | 17 | 2045 | ||
| GenBank mRNA | AK046740.1 | - | 17 | 2015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc7b| Gene Symbol: Ttc7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033213.1 | NP_001028385.1 | 17 | 2045 | ||
| GenBank mRNA | AK046740.1 | - | 17 | 2015 | ||