siRNA Details
| Gene Symbol: Sptssa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134054.2 | NP_598815.2 | 2 | 241 | ||
| GenBank mRNA | AK003171.1 | - | 2 | 178 | ||
| AK003275.1 | - | 2 | 182 | |||
| AK008904.1 | - | 2 | 198 | |||
| AK018380.1 | - | 2 | 218 | |||
| AK137543.1 | - | 2 | 216 | |||
| BC022674.1 | - | 2 | 167 | 4236922 | ||
| BY072251.1 | - | 2 | 242 | |||

Sptssa
Genomic Map