siRNA Details
| Gene Symbol: E030019B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053157.1 | - | 5 | 2141 | ||
| AK138262.1 | - | 2109 | ||||
| BC064451.1 | - | 7 | 1570 | |||
| KF856953.1 | - | 8 | 1765 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E030019B13Rik| Gene Symbol: E030019B13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK053157.1 | - | 5 | 2141 | ||
| AK138262.1 | - | 2109 | ||||
| BC064451.1 | - | 7 | 1570 | |||
| KF856953.1 | - | 8 | 1765 | |||