siRNA Details
| Gene Symbol: Tecpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081057.2 | NP_001074526.1 | 8 | 1253 | ||
| NM_001289510.1 | NP_001276439.1 | 7 | 1201 | |||
| NM_175336.3 | NP_780545.2 | 8 | 1253 | |||
| GenBank mRNA | AK031736.1 | - | 7 | 1158 | ||
| AK129113.1 | - | 7 | 1205 | |||
| AK147592.1 | - | 7 | 1201 | |||
| BC041778.1 | - | 5 | 845 | |||
| BC138354.1 | - | 7 | 1196 | 8861374 | ||
| BC145250.1 | - | 7 | 1196 | |||

Tecpr2
Genomic Map