siRNA Details
| Gene Symbol: Slc25a47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012310.2 | NP_001012310.2 | 4 | 244 | ||
| GenBank mRNA | AK149369.1 | - | 4 | 250 | ||
| BC035527.1 | - | 4 | 245 | |||
| BC042739.1 | - | 3 | 372 | |||
| BC055027.1 | - | 4 | 237 | |||
| BC069939.1 | - | 4 | 233 | 5053384 | ||
| BC089616.1 | - | 4 | 244 | 6760348 | ||

Slc25a47
Genomic Map